PIG® Oil-Only Absorbent Mat Pad in Dispenser Box, New Pig
Supplier: New Pig
MAT485
89094-960CS 109.7 USD
89094-960
PIG® Oil-Only Absorbent Mat Pad in Dispenser Box, New Pig
Spill Control Materials
Eight layers of 100% polypropylene are thermally bonded to give PIG Mat excellent strength; won't rip, tear or fray even when saturated
Can be wrung out and incinerated after use to reduce waste or for fuels blending. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Dispenser box makes mat easy to access, carry and store while protecting contents from dirt and moisture. Pads are ideal for catching drips and soaking up spills. Easy-tear perforations let you take only what you need, so you use less mat and save money.
Lightweight construction is economical for small tasks and cleanups.
Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave behind liquids or fiber residue. Absorbs and retains oils and oil-based liquids, including lubricants and fuels, without taking in a drop of water. Bright white color makes absorbed oil easier to see; draws attention to machine leaks and clearly shows saturation level during spill response. Floats to clean up spills on water.