VWR® Traceable® Expanded Range Conductivity Meter
Supplier: VWR International
93676-73
89094-958EA 838.14 USD
89094-958
VWR® Traceable® Expanded Range Conductivity Meter
Electrochemistry Meters Conductivity Meters
Accurate answers in five seconds with a meter that automatically selects proper range and displays the exact answer.
  • Compatible with all accreditation analysis requirements
  • Accurate
  • Automatically selects power range
Ideal for checking water purity in stills, deionizers, reverse osmosis, measuring total dissolved solids and making solutions.
Compatible with all accreditation analysis requirements - perfect for labs wanting to maintain accreditation.
Units fulfills requirements for CAP, ASTM, NCCLS, CLSI, ACS, CLIA, AOAC, EPA, APHA, AWWA, WEF, USGS, USP, ISO and Federal and State regulations.
Certifications: Traceable to NIST for accuracy. To assure accuracy an individually serial-numbered Traceable® Certificate is provided from VWR® ISO/IEC 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Standards and Technology).
Ordering information: Supplied with ISO 17025 Traceable® Certificate, epoxy probe, batteries, carrying case and computer output capabilities.