VWR® Traceable® Waterproof Food HACCP Thermometer
Supplier: VWR International
89094-774 10171-318 10171-482 10171-598
89094-774EA 300.12 USD
89094-774 10171-318 10171-482 10171-598
VWR® Traceable® Waterproof Food HACCP Thermometer
Thermometers Digital Thermometers
This unit combines a non-invasive infrared thermometer with a piercing stainless steel probe thermometer.
  • Traceable
  • Temperature in °C and °F
  • Stainless steel probe and no-contact infrared option
  • LED indicators for HACCP zones
Three LED indicators for HACCP zones: under 4°C (40°F), between 4 to 60°C (40 to 140°F), and over 60°C (140°F)
A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Easy-to-use membrane switches and rugged ABS body make this thermometer ideal for food labs, food processing, food preparation, and brewing. Waterproof construction allows use in wash-down and other food areas. Displays current temperature, minimum or maximum temperature, low battery, and emissivity value. Readings are updated every second. Large, bright LCD digits are 6mm (1/4).
Ordering information: Supplied with battery.