VWR® Traceable® Pen for Salinity
Supplier: VWR International
89094-766
89094-766EA 175.58 USD
89094-766
VWR® Traceable® Pen for Salinity
Electrochemistry Meters Salinity Meters
Unit has an instant-response probe and solid-state thermistor for quick, precise readings.
  • Traceable®
  • Temperature in °C and °F
  • Tough ABS plastic housing
  • Water-resistant design
  • Memory function retains calibration data even when turned off
  • Fast setting to calibration standards
Tough ABS plastic housing and water-resistant design keep out moisture, dust, and dirt, assuring long life in severe laboratory or harsh plant environments.
Meter fulfills requirements for CAP, ASTM, NCCLS, CLSI, ACS, CLIA, AOAC, EPA APHA, AWWA, WEF, USGS, USP, ISO, and federal and state regulations. A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Easy-to-read LCD digits are 1.3 cm (¹/₂") high. Features include arrow keys fast setting to calibration standards, temperature compensation, and programmable temperature coefficient. Calibration data is stored even when unit is turned off.