VWR® Traceable® Waterproof Type-K Thermometer
Supplier: VWR International
89094-738 10171-320 10171-484 10171-600
89094-738EA 254.37 USD
89094-738 10171-320 10171-484 10171-600
VWR® Traceable® Waterproof Type-K Thermometer
Thermometers Digital Thermometers
Wide-range thermometer has shock-resistant ABS plastic casing.
  • Traceable
  • Temperature in °C and °F
  • Waterproof design
  • Highly visible display
  • Minimum and maximum temperature recall
Waterproof design stops dirt, dust, fumes, and water, making it ideal in harsh lab or plant environments. Features include °C or °F display, minimum and maximum temperature recall, differential temperature from a set point, hold button to capture current reading, and backlight button. The microprocessor updates the display 2.5 times per second. The 1.6 cm (5/8") digits are readable from 3m (10').
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with 9-volt battery, carrying case, fast-response type-K beaded probe with 1.2 m (4') cable, and waterproof rubber cover for probe connector. For other available type-K probes (sold separately), see the 61161-372 series.