VWR® Traceable® Micro-Anemometer with Thermometer
Supplier: VWR International
89094-736 10754-706
89094-736EA 187.37 USD
89094-736 10754-706
VWR® Traceable® Micro-Anemometer with Thermometer
Anemometers
Compact anemometer displays air velocity in meters per second, feet per second, miles per hour, kilometers per hour, and knots.
  • Traceable®
  • Displays air velocity, max. air velocity measured, temperature, wind chill temperature
  • Tough ABS plastic housing
  • Temperature in °C and °F
Unit also displays maximum air velocity measured, temperature, and wind chill temperature. Temperature is displayed in °C and °F. Measurements are updated twice per second. Tough ABS plastic housing and water-resistant design keep out moisture, dust, and dirt, assuring long life in severe lab or harsh plant environments. Features include intuitive one-button operation and electroluminescent backlight. High-contrast LCD digits are 1 cm (3/8") high.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.