VWR® TraceClean® Containers with Chemical Preservative
Supplier: VWR International
89094-186 89094-188 89094-200
89094-186CS 101.91 USD
89094-186 89094-188 89094-200
VWR® TraceClean® Containers with Chemical Preservative
Bottles Water Sampling Bottles
Containers are filled with chemical preservative to maintain sample integrity until analysis can be performed.
  • Containers are made of HDPE or glass
  • Provided with polypropylene cap
  • Cap may or may not have a liner
  • Hydrochloric acid and sulfuric acid are for oil
HDPE containers are for water samples. Bottle 89094-186 and packer 89094-188 are supplied with polyfoam-lined polypropylene cap, and NALGENE® jars are supplied with linerless polypropylene cap. Glass jars are supplied with polypropylene cap with 3.2 mm (0.125") fluoropolymer liner.