CinchStrap-EG™ Grommetted Cable Management Straps, Rip-Tie®
Supplier: Rip-Tie, Inc.
N-07-G10-BK N-10-G02-BK N-07-G02-BK N-10-G10-BK O-16-G1P-BK
89093-542PK 74.58 USD
89093-542 89093-544 89093-540 89093-546 89093-548
CinchStrap-EG™ Grommetted Cable Management Straps, Rip-Tie®
Clamps General Purpose Clamps
Professional quality, heavy-duty straps can be attached to wiring racks, mobile carts, walls, or doorways to provide simple cable or wiring management and organization
Any fastener up to 0.64cm (1/4") can be used for mounting. Non-magnetic brass grommets spread the fastener load evenly and eliminate the risk of ripping fabric. Durable straps are reusable for up to 10,000 refastenings, and can be oriented in any direction. Color: black.