SP Bel-Art Rack Slanter, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18890-0100
89093-458EA 40.27 USD
89093-458
SP Bel-Art Rack Slanter, Bel-Art Products, a part of SP
Racks Tube Racks
Tilt test tube racks to 25°, 35°, or 45°.
  • Adjustable angle
  • Ideal for creating agar slant tubes
  • Built-in magnifier aids in viewing Petri dishes
  • Autoclavable
Adjusting the angle of racks reduces hand strain when working with pipettors
Base legs have predetermined slots that the stand slides in and out of to easily change angles. Fits racks up to 29.3×24.2×8.3 cm (119/16×99/16×31/4"). Steam autoclavable at 121 °C (250 °F).
Also ideal for creating agar slant tubes or when using the Mini-Magnifier (54848-306) to aid in the viewing of Petri dishes.