SP Bel-Art Poxygrid® Centrifuge Tube Rack, 500 ml, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F19856-0500
89093-450EA 49.24 USD
89093-450
SP Bel-Art Poxygrid® Centrifuge Tube Rack, 500 ml, Bel-Art Products, a part of SP
Racks Tube Racks
The only wire rack that holds 500 mL conical centrifuge tubes.
  • Epoxy-coated steel wire construction
  • Conical-bottomed centrifuge tubes
  • Autoclavable
Support, transport, and store up to four 500 mL conical bottom centrifuge tubes. Epoxy-coated steel wire. Steam autoclavable at 121ºC (250ºF).