SP Bel-Art Conical Tube Holder, 15 and 50 ml, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18796-0000
89093-448PK 21.51 USD
89093-448
SP Bel-Art Conical Tube Holder, 15 and 50 ml, Bel-Art Products, a part of SP
Racks Tube Racks
The most versatile tube holder on the market, this set-up takes just seconds and gives users the choice of creating a 15 mL or a 50 mL tube holder.
  • Easy set-up
  • Open uprights allow clear visibility
  • Conical bottom
  • Easy to remove from holder
  • Polypropylene construction
  • Autoclavable
Unlike other tube holders, the open uprights allow users to view the entire contents of the tube
Tubes can be easily lifted from the holder, or pinch the uprights to carry the holder with the tube. Tube holders are manufactured from autoclavable polypropylene.
No more guessing if there is liquid remaining in the conical bottom.