Axygen® Pre-Sterilized Individually Wrapped Blue Pestle, Corning
Supplier: Corning
PES-15-B-SI
89093-446CS 512.82 USD
89093-446
Axygen® Pre-Sterilized Individually Wrapped Blue Pestle, Corning
Mortars and Pestles Pestles
Durable and inert polypropylene construction
  • Designed to fit 1.5mL and 2.0mL centrifuge tubes
  • Individually wrapped and sterilized