59-050 Liquid Protection Sleeve, Neoprene, Ansell
Supplier: Ansell Healthcare
950235
89093-016CS 1052 USD
89093-016
59-050 Liquid Protection Sleeve, Neoprene, Ansell
Workwear
With elastic at both ends, this neoprene sleeve provides excellent protection against chemicals, snags, punctures, and abrasions.
  • Excellent resistance to a wide range of chemicals
  • Extremely flexible and lightweight for level of protection
  • Durable and cost-effective
  • Ambidextrous
Use in the chemical processing, electronics manufacturing, food processing, food service, forestry, natural gas, oil refining, and pharmaceutical industries. Uses include chemical processing and industrial maintenance.