Biohazard Wipes and Liners, Current Technologies
Supplier: Current Technologies
BH16100 BH33400 BH44985 BH90044 BH91033 BH92910 BH93016 BH 16016 ET BH 10600 BH32000 BH91625 BH44200 BH11016ET
16200-204CS 458.6 USD
16200-204 16200-206 16200-208 16200-210 16200-212 16200-214 16200-216 89092-532 89092-534 82030-864 10147-840 10147-842 10147-844 10147-848
Biohazard Wipes and Liners, Current Technologies
Wipes Disposable Wipes and Towels Disinfectant Wipes
Low-linting wipes and liners are specifically designed to protect workers and labs from contamination from blood, body fluids, tissue, specimens plus chemicals.
Wipes and liners are available in several sizes and in large rolls for cutting to size. Wipes are also available in standard absorbency, which can absorb 9× its own weight, heavy absorbency, which can absorb 12× its weight and super absorbency, which can absorb 25× its weight.
Specifically sized dispensers are constructed of clear acrylic.
Absorbent white side soaks up blood, tissue, specimen, body fluids, and chemicals. Bright orange barrier side, made of coextruded plastic, blocks soak-through to gloves, hands and counters, limiting the spread of contamination. Wipes are impermeable to strong solvents including xylene and xylene substitutes, acetone, ketones, alcohols, and formalin. Use to pop caps, wipe pipets and probes, handle specimens and line benches, counters and trays, clean up spills.
Ordering information: Bio-Screen wipes and liners are non-sterile and are not for use on patients.