Accessories for PYREX® Continuous Liquid/Liquid Extractor, Heavier Than Water, One Piece, Corning
Supplier: Corning
3920-MBO
89091-652CS 642.75 USD
89091-652
Accessories for PYREX® Continuous Liquid/Liquid Extractor, Heavier Than Water, One Piece, Corning
Extractors Liquid-Liquid Extractors
Made of PYREX® borosilicate glass. These continuous extractor systems are for use with solvents of heavier specific gravity than sample liquid being extracted.
  • Extraction can be performed outside the hood
  • Designed to meet EPA test method requirements outlined by SW-846, RCRA, and CLP
The sidearm is sealed to the extractor body just below the 45/50 [ST] outer joint. The sidearm is sealed higher on the body to reduce the chance of water spilling over while draining remaining solvent from the extractor body at the end of the extraction. Extraction can be performed outside the hood. However, as dictated by safe laboratory practices, it is recommended that some type of ventilation system be used above or near the units in case of a drop in condenser water temperature or loss of condenser cold water supply.
Delivery information: Supplied with Allihn condenser with 45/50 [ST] inner joint and 500m; PYREX® round bottom boiling flask with 24/40 [ST] outer joint.