PYREX® Reducing Adapters with [ST] Joints, Corning
Supplier: Corning
8800-2419 8800-2924 8800-3424
89091-642CS 616.27 USD
89091-642 89091-644 62900-088
PYREX® Reducing Adapters with [ST] Joints, Corning
Adapters Adapters, Conical Ground Joints
PYREX® glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.