PYREX® Heavy Wall Drying Tray, Corning
Supplier: Corning
3170-12
89091-600CS 1815.35 USD
89091-600
PYREX® Heavy Wall Drying Tray, Corning
Trays/Dishes/Pots Drying Trays
These heavy wall dishes made of PYREX® borosilicate glass will withstand hot air or steam sterilization and are useful for drying, staining or developing trays.
  • Excellent chemical resistance
  • Good temperature resistance
  • Easy to clean
They can be used with gel electrophoresis or paper chromatography.
Caution: For oven use only. Do not use over direct heat.