PYREX® Distilling/Suction Adapter with [ST] Joints, Corning
Supplier: Corning
9420-24
89091-574CS 941.4 USD
89091-574
PYREX® Distilling/Suction Adapter with [ST] Joints, Corning
Distillation Components Distillation Adapters
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
These distilling suction tube adapters convert ordinary boiling flasks with 24/40 standard taper jointed necks to vacuum-type standard taper receivers. They have a 24/40 standard taper outer joint at the top and a 24/40 standard taper inner joint at the bottom. The sidearm is approximately 10 mm Ø by 25 mm long.