PYREX® Centrifuge Tubes with [ST] Stopper, Conical-Bottom, without Graduations, Corning
Supplier: Corning
8424-50
89091-458CS 633.1 USD
89091-458
PYREX® Centrifuge Tubes with [ST] Stopper, Conical-Bottom, without Graduations, Corning
Tubes Centrifuge Tubes
PYREX®, borosilicate glass, round bottom, with ground joint.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These PYREX® round bottom, non-graduated centrifuge tubes have a No. 13 standard taper solid glass pennyhead stopper.Rockefeller Institute of Medical Research type.
Reusable and non-sterile.