PYREX® Volumetric Pipettes, Color-Coded, Class A, Corning
Supplier: Corning
7100-1 7100-3 7100-4 7100-10 7100-20 7100-25 7100-100 7100-50 7100-2 7100-5 7100-15
53044-037CS 243.85 USD
53044-037 53044-070 53044-092 53044-139 53044-172 53044-194 89091-078 89091-456 53044-059 53044-117 53044-140
PYREX® Volumetric Pipettes, Color-Coded, Class A, Corning
Pipettes Graduated Pipettes
Colored markings remain sharply legible for the life of pipet.
  • Reusable, rugged construction
  • Color-coded
  • Colored enamel graduations and markings
Ruggedly constructed and properly proportioned, pipettes have accuracy tolerances to meet or exceed the strict requirements for Class A Volumetric Ware as indicated by ASTM E969.