PYREX® Boiling Flasks, Florence, Flat Bottom, Long Neck, with Standard Taper Joint, Corning
Supplier: Corning
4080-250
89091-254CS 662.92 USD
89091-254 89091-418
PYREX® Boiling Flasks, Florence, Flat Bottom, Long Neck, with Standard Taper Joint, Corning
Flasks Flat Bottom Flasks
PYREX® borosilicate glass, long neck.
  • High thermal shock resistance
  • Provide contamination free results
  • High chemical durability
These flasks have full length outer standard tapered 24/40 joint. The normal working temperature is 230 °C (446 °F).