PYREX® Heavy-Wall Centrifuge Bottle with Screw Cap, Corning
Supplier: Corning
1261-200
89091-232CS 1345.91 USD
89091-232
PYREX® Heavy-Wall Centrifuge Bottle with Screw Cap, Corning
Bottles Centrifuge Bottles
PYREX® borosilicate glass, with PP screw cap.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Cylindrical, wide mouth, bottle is ideal for sputum specimen collection. Digestion, shaking, neutralizing, and centrifuging can be performed in one container. Supplied with tight-fitting, white polypropylene screw cap with fluoropolymer resin coated liner.