PYREX® Gas Oil Distillation Percent Scale Graduated Cylinders, Serialized and Certified, Class A, To Contain, Corning
Supplier: Corning
3065-200
89091-230CS 1257.82 USD
89091-230
PYREX® Gas Oil Distillation Percent Scale Graduated Cylinders, Serialized and Certified, Class A, To Contain, Corning
Cylinders Measuring Cylinders
Borosilicate glass.
  • Calibrated to contain TC in percent
  • Graduations are in durable white enamel
  • Used to measure the distillate in ASTM D86
Delivery information: Supplied with bumper guards.