PYREX® Blunt Tip Imhoff Sediment Cone, Corning
Supplier: Corning
2180-439
89091-192CS 2048.93 USD
89091-192
PYREX® Blunt Tip Imhoff Sediment Cone, Corning
Funnels Sedimentation Cones
Made of PYREX® borosilicate glass.
  • Also graduated and marked at 1000 ml
  • Blunt tip increases ruggedness and cleaning ease
  • Very good chemical and temperature resistance
These blunt tip cones are used with heavy sediments, where ability to read small volumes is unimportant. Graduated from 0 ml to 1 ml in 0.1 ml increments, 1 to 10 ml in 0.5 ml increments and 10 ml to 40 ml in 1 ml increments.