PYREX® Conical Centrifuge Tubes with White Graduations, Beaded Rim, Corning
Supplier: Corning
8080-15 8080-50
89091-174CS 345.65 USD
89091-174 21054-021
PYREX® Conical Centrifuge Tubes with White Graduations, Beaded Rim, Corning
Tubes Centrifuge Tubes
PYREX® borosilicate glass.
These tubes have white enamel graduations. The 10 and 15 ml sizes are graduated upward from tip in 0.1 ml increments. The 50 ml size is graduated upward from tip to 10 ml in 0.5 ml increments, and from 10 to 50 ml in 1 ml increments. Conical bottom with beaded rim.