PYREX® Squibb Separatory Funnel, PTFE Stopcock, Standard Taper Joints, Corning
Supplier: Corning
6413-125
89091-140CS 911.02 USD
89091-140
PYREX® Squibb Separatory Funnel, PTFE Stopcock, Standard Taper Joints, Corning
Funnels Separating Funnels
These PYREX® glass separatory funnels have Polytetrafluorethylene (PTFE) No
2 standard stopcocks and inner and outer 19/22 Standard Taper joints. The PTFE plug fits into the micro-finish barrel. The plug also reduces freezing problems and grease contamination.