PYREX® Dean Stark Distallation Receiver, [ST] Joints, Corning
Supplier: Corning
3602-10
89091-104CS 936.08 USD
89091-104
PYREX® Dean Stark Distallation Receiver, [ST] Joints, Corning
Distillation Components Distillation Receivers
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
This 10 ml Dean Stark distilling receiver is graduated from 0 to 1 ml in 0.1 ml intervals, with a maximum scale error of 0.05 ml. The graduation intervals from 1.0 to 10.0 ml are 0.2 ml, with a maximum scale error of 0.1 ml.
Certifications: ASTM E-123