PYREX® Oil Pear-Shaped Centrifuge Tube Lifetime-Red™ with Graduations, Corning
Supplier: Corning
8200-100
89091-076CS 2135.56 USD
89091-076
PYREX® Oil Pear-Shaped Centrifuge Tube Lifetime-Red™ with Graduations, Corning
Tubes Centrifuge Tubes
Made of PYREX® borosilicate glass.
  • Scale and figures are durable white enamel
  • Red color-band
  • Narrow mouth
  • Beaded rim
These pear-shaped centrifuge tubes are used for the determination of water and sediment in petroleum products. They are fabricated from mold-blown blanks to insure uniform shape and greater mechanical strength.
The stem is graduated from 0 to 1.5 ml in 0.1 ml divisions. The body is graduated from 1.5 to 5 ml in 0,5 ml divisions; from 5 to 10 ml in 1 ml divisions; and at 15, 20, 25, 50 and 100 ml.
Certifications: Conform to ASTM D-96 and D-1966.