Corning® Low-Profile Magnetic Stirrers, 230 V
Supplier: Corning
6796-210
89090-990CS 733.52 USD
89090-990
Corning® Low-Profile Magnetic Stirrers, 230 V
Stirrers Magnetic Stirrers
The Corning® stirrers have a durable glass-ceramic 4×5" Pyroceram® top and offer superior performance and reliability in a low profile, space-saving design.
  • 230 V model
  • Designed with CEE7-7 plug
  • Microprocessor controlled to automatically adjust motor strength to any solution
  • Enclosed electronics design protects components, improves performance, and prolongs product life
  • Two piece casting design deflects spills away from electronics
  • Built in mount for an optional support rod
Exclusive closed-loop stirring control monitors and regulates the stirring speed for aqueous, viscous, or semi-solid solutions. A stronger, larger magnet improves stir bar coupling and stirring capability.
Delivery information: Includes a built in mount for an optional support rod and 1×9.5 stir bar. Manufacturer's two-year warranty on parts and labor. Instruction manual, technical information book, and three-wire cord and plug.
Caution: 230 V model. Corresponding models with voltage suitable for use in US (89090-986, 89090-988) are also available, please see separate product page.