PYREX® Separatory Funnels, Squibb, Pear-Shape, Fluoropolymer Resin Stopcock, Corning
Supplier: Corning
6404-1L 6404-2L 6404-60 6404-125 6404-250 6404-500
30356-700CS 345.47 USD
30356-700 30356-722 89091-508 89090-654 89090-792 89090-880
PYREX® Separatory Funnels, Squibb, Pear-Shape, Fluoropolymer Resin Stopcock, Corning
Funnels Separating Funnels
PYREX® Borosilicate glass.
With PTFE stopcock and linear, high-density, closed-bottom polyethylene stoppers which conform to [ST] stopper dimensions.