PYREX® Volumetric Flasks, Lifetime Red™, Graduated, Snap Cap, Class A, Corning
Supplier: Corning
5600-25 5600-100 5600-250 5600-1L 5600-500 5600-2L 5600-50
29610-285CS 502.35 USD
29610-285 89090-598 89090-672 89090-688 89090-706 89090-744 89090-838 89090-904
PYREX® Volumetric Flasks, Lifetime Red™, Graduated, Snap Cap, Class A, Corning
Flasks Volumetric Flasks
PYREX® borosilicate glass, clear, snap cap.
These flasks have the capacity mark etched as a fine line through a narrow red band into clear glass.
Certifications: ASTM E-694, E-542, E-288.