PYREX® Frierichs Condenser, Corning
Supplier: Corning
2640-350
89090-520CS 1551.27 USD
89090-520
PYREX® Frierichs Condenser, Corning
Condensers
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
This PYREX® Friedrichs type condenser affords very efficient operation. The helical inner tube fits closely within the jacket. The vapor tube is sealed to the jacket at an angle of 75° and has an outer 24/40 Standard Taper joint.