PYREX® Barrett Distilling Apparatus, with Stopcock and 24/40 Standard Taper Joints, Corning
Supplier: Corning
3622-20
89090-486CS 1726.89 USD
89090-486
PYREX® Barrett Distilling Apparatus, with Stopcock and 24/40 Standard Taper Joints, Corning
Distillation Components Distillation Receivers
This PYREX® distilling apparatus is designed for determining water content in petroleum or bituminous products.
  • With stopcock (glass plug) and 24/40 [ST] joints
  • Very good temperature resistance
  • Very good chemical resistance
It is graduated upwards from the stopcock to 3 ml, in 0.2 ml divisions, and from 3 ml to 20 ml in 0.5 ml divisions.
The apparatus can also can be used as Dean Stark distillation apparatus specified in British Standard 756, but utilizes a common U.S. ground joint. It is also an optional setup for ASTM D-95.