PYREX® VISTA™ Rimless Culture Tubes, Corning
Supplier: Corning
70820-6 70820-16X 70820-16XX 70820-22 70820-25
89090-296CS 82.16 USD
89090-296 89090-320 89090-336 89090-376 89090-400 89090-404 89090-410 89090-414
PYREX® VISTA™ Rimless Culture Tubes, Corning
Tubes Culture Tubes
Borosilicate glass.
Reusable rimless culture tubes offer greater convenience in plugging and rack storage. Round bottom tubes feature fire polished ends. All tubes have a large blue enamel marking spot for simple identification, with the exception of 50, 55 and 70 ml sizes.
Delivery information: Supplied without caps.