Corning® Polypropylene Cap with 2 Stainless Steel Tubing Ports, Corning
Supplier: Corning
401654
89089-806CS 272.52 USD
89089-806
Corning® Polypropylene Cap with 2 Stainless Steel Tubing Ports, Corning
Closures Bottle Closures
Autoclavable polypropylene caps feature two 15.2 cm (6") stainless steel tubes
Caps fit Corning glass and plastic vessels with GL45 mm threads. Designed for bioprocess and scale-up applications.