Corning® PBT GL 32 Caps, Corning
Supplier: Corning
1395-32HTC
89089-690CS 125.72 USD
89089-690
Corning® PBT GL 32 Caps, Corning
Closures Test/Sample Tube Closures
The red Corning® polybutylene terephthalate (PBT) high temperature caps have a GL32 thread, and can be dry heat sterilized at up to 180 °C
Caps have a Polytetrafluorethylene (PTFE) liner for efficient sealing.