Sodium acetate trihydrate ≥99%, granular USP suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Acetic acid sodium salt trihydrate

SO105-100KGB SO105-12KG SO105-100KG SO105-500GM SO105-2.5KG SO105-50KGBL
700004-370EA 2157.25 USD
700004-370 89050-448 89089-164 89050-452 89050-454 89050-450
Sodium acetate trihydrate ≥99%, granular USP suitable for use as excipient
Sodium acetate trihydrate

Sodium Acetate, Trihydrate, Granular, USP is a moderately water soluble crystalline source of Sodium and serves as processing aid excipient. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: H₃CCOONa·3H₂O
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR