SP Bel-Art Cover for Glass Disposal Carton, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F13204-0001
89088-050CS 411.06 USD
89088-050
SP Bel-Art Cover for Glass Disposal Carton, Bel-Art Products, a part of SP
Boxes Waste Disposal Boxes
Dishwasher-safe and autoclave-safe, the reusable Disposal Carton Cover opens quickly and easily by pressing the built-in release tab.
  • Reusable design
  • Dishwasher-safe
  • Autoclavable
  • Wider opening for safer and more sanitary deposit of wastes
There are no batteries required. The cover fits any disposal carton with a 31.5x31.5 cm (12½x12½") opening (including the SCIENCEWARE® Glass Disposal Floor Standing Carton), and features a smooth 10.2x10.2 cm (4x4") area for applying labels. The center panel is green to match the printing on the glass disposalMore Product Information