SP Wilmad-LabGlass PTFE Thermometer Adapter with Compression Cap, SP Industries
Supplier: SP Industries
LG-1453-100
89087-526EA 50.08 USD
89087-526
SP Wilmad-LabGlass PTFE Thermometer Adapter with Compression Cap, SP Industries
Adapters Adapters, Conical Ground Joints
This inner joint thermometer adapter has a threaded compression cap, four saddle O-Rings, and one Viton® O-Ring for use with plain stem thermometers and inlet tubes.
  • Threaded compression cap, four saddle O-Rings, and one Viton® O-Ring included
  • Accommodates thermometers and inlet tubes
  • Made of PTFE
The adapter accepts thermometers with outer diameters from 3 to 7mm.