Orthophosphoric acid ≥85% ACS

Supplier: Spectrum Chemical Mfg. Corp.
Danger

P1095-1LT P1095-100ML P1095-2.5LTG P1095-2.5LTP P1095-500MLG P1095-2.5LTP P1095-200LTB P1095-2.5LTG P1095-200LTB
89087-354EA 291.04 USD
89087-354 89087-350 700000-608 700000-610 89087-352 89087-358 700000-612 89087-356 89087-362
Orthophosphoric acid ≥85% ACS
Orthophosphoric acid

Phosphoric Acid, Reagent, ACS is a also known under the name othophosphoric acid. A mineral acid that is most often presented as a non-volatile and clear liquid that a viscous texture. It has a wide field of uses, such as a reagent, most commonly found in a liquid solution of 75% to 85%, it can also be employed to remove rust from metal surfaces, used as a food additive under the name E338 used in many soft drinks and in orthodontics as an etching solution. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness

Formula: H₃PO₄
Melting Pt: 28 °C
Density: 1.685
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Assay 85% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR