VWR® Sterile Sample Bags
Supplier: VWR International
89000-644 89000-190 82007-696 82007-698 82007-700 82007-706 82007-708 82007-714 82007-716 82007-718 82007-724 82007-726 82007-728 83007-556 83007-558 83007-560 83007-562 89000-636 89000-638 89000-640 89000-642 89000-648 89000-650 89004-418 89004-420 89004-422 89004-424 89085-528 89085-530 89085-532 89085-538 89085-540 89085-542 89085-544 89085-546 89085-548 89085-550 89085-552 89085-554 89085-556 89085-558 89085-560 89085-580 10818-386 89085-535 89085-537 76196-370 76196-372 76196-376 76214-710 76221-334 76265-604 76289-492 11216-104
89000-644CS 403.18 USD
89000-644 89000-190 82007-696 82007-698 82007-700 82007-706 82007-708 82007-714 82007-716 82007-718 82007-724 82007-726 82007-728 83007-556 83007-558 83007-560 83007-562 89000-636 89000-638 89000-640 89000-642 89000-648 89000-650 89004-418 89004-420 89004-422 89004-424 89085-528 89085-530 89085-532 89085-538 89085-540 89085-542 89085-544 89085-546 89085-548 89085-550 89085-552 89085-554 89085-556 89085-558 89085-560 89085-580 10818-386 89085-535 89085-537 76196-370 76196-372 76196-376 76214-710 76221-334 76265-604 76289-492 11216-104
VWR® Sterile Sample Bags
Bags Sample Bags
These sample bags are ideal for transportation and storage of solids, semisolids, and liquids for environmental and carcass sampling, biomedical and pharmaceutical research, quality assurance procedures, food industry applications, and clinical and veterinary medicine.
  • Pliant polyethylene sterile tubing
  • Highly resistant
  • Wire closure for an airtight seal
  • Round wire bags have two wires for closure
Bags are made of pliant, highly resistant virgin polyethylene sterile tubing with no side seals. They feature a wire closure for an airtight seal, and wide-sealed bottoms for added leakproof protection. To use, tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal.
Note: Do not use with alcoholic substances or where temperature exceeds 100 °C (212 °F).
Note: Do not use with alcoholic substances or where temperature exceeds 100°C (212°F).
Certifications: Comply with EPA, FDA, USDA, and HACCP regulatory and industry requirements.
Ordering information: Bags 89085-560 and 76289-492 are packaged in a double pouch for cleanroom applications.