VWR® Basic Frosted Microscope Slide without Tissue Paper
Supplier: VWR International
0302-2222
89085-399CS 293.01 USD
89085-399
VWR® Basic Frosted Microscope Slide without Tissue Paper
Slides Microscope Slides
VWR® Frosted Microscope Slides without Tissue Paper are specially designed with consistent and clean surfaces for staining experiments.
  • Consistent, clean surfaces
  • Frosted edge for marking
They are ideal for histology, hematology, and microbiology applications. Slides feature frosted areas on both sides of one end for convenient marking. A fine polished edge and 45° clipped corners promote ease and safety in handling. Slides are packaged 72 per pack, 20 packs per case.