D-(+)-Biotin ≥97.5%, powder USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
B1103-5GM
B1103-1KGBL
B1103100GMBL
B1103-1GM
B1103-25GM
B1103-500MG
B1103-500GMBL
89085-276EA
1080.01
USD
89085-276
700009-418
89085-280
89085-274
89085-278
89085-272
75810-670
D-(+)-Biotin ≥97.5%, powder USP
D(+)-Biotin
Biotin, Powder, USP is a water-soluble B vitamin.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₁₀H₁₆N₂O₃S MW: 244.31 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00005541 CAS Number: 58-85-5 Merck Index: 13,01231 |
Specification Test Results
Assay | 97.5 - 102.0 % |
Specific Rotation, [A]D25 | +89° to +93° |
Related Compounds | To Pass Test |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...