Separatory Globe Funnel, Ace Glass
Supplier: Ace Glass
7221-09
89085-040EA 204.04 USD
89085-040
Separatory Globe Funnel, Ace Glass
Funnels Separating Funnels
This Globe-Style Separatory Funnel features a 175 mm stem below the PTFE stopcock and is supplied with a flask length top stopper.