Bulb Condenser, Reflux, Ace Glass Incorporate
Supplier: Ace Glass
6042-04 6042-06 6042-10 6042-08 6042-02 6042-114 6042-124
89084-960EA 443.13 USD
89084-960 89214-970 89214-972 89235-624 10099-080 10099-082 10099-084
Bulb Condenser, Reflux, Ace Glass Incorporate
Condensers
This new style, compact, high-output reflux condenser has an inner double wall and thimble-shape internal bulb
Overall height is approximately 210 mm, but the output is equal to condensers with twice the length. Unit has standard taper reinforced outer or cone top joint and standard taper drip-tip inner or drip-tip socket bottom joint. Top and bottom hose connections size D, for 3/8" tubing. Fabricated from borosilicate glass. 