Spiral Coil Condenser, Reflux, Ace Glass
Supplier: Ace Glass
6040-02 6040-04
89084-956EA 326.48 USD
89084-956 89214-974
Spiral Coil Condenser, Reflux, Ace Glass
Condensers
This compact spiral coil style condenser has a tightly wrapped coil for maximum cooling
Excellent for use with high vapor pressure solvents. Jacket length is approximately 100 mm, and overall height is approximately 210 mm. Features a 75° angle top standard taper outer joint, and standard taper vertical inner bottom joint. Top and bottom hose connections size C, for 9.5 mm (3/8") or 7.9 mm (5/16") I.D. tubing. Fabricated from borosilicate glass.