Clamp Holder, All-Position, Ace Glass
Supplier: Ace Glass
11090-17
89084-748EA 128.21 USD
89084-748
Clamp Holder, All-Position, Ace Glass
Clamps General Purpose Clamps
This all-position holder connects at any angle, in any plane
The apparatus consists of two holders connected by a double plate-joint which sets the holders 90° from each other. Each holder can be rotated 360°. Grips rods up to 19mm diameter.