Labjaws FootPlate, Ace Glass Incorporated
Supplier: Ace Glass
11174-13
89084-716EA 52.02 USD
89084-716
Labjaws FootPlate, Ace Glass Incorporated
Support Stands
The Labjaws Footplate holds Flexaframe poles
It secures them to floors, walls and tables with screws. Mount lattice permanently.