Support Stand for Bench-Scale Chromatography Columns, Ace Glass Incorporated
Supplier: Ace Glass
5868-122
89084-674EA 1292.81 USD
89084-674
Support Stand for Bench-Scale Chromatography Columns, Ace Glass Incorporated
Support Stands
This single-post stand is designed for 30.5 cm (12") and 45.7 cm (18") height chromatography columns
It has an “H” shaped base and a single stainless steel bar to provide great stability for the columns.
Delivery information: The stand includes base, rod, base plate, clamp holder, and chain clamp.