Clamp, Utility, 3-Prong, Ace Glass
Supplier: Ace Glass
11062-14
89083-216EA 103.53 USD
89083-216
Clamp, Utility, 3-Prong, Ace Glass
Clamps General Purpose Clamps
This three-prong grip clamp has jaws and holder made of zinc alloy
The clamp can be extended in length to 16mm, and firmly locked in any position by means of the provided wing-nut. It will hold objects up to 57mm in diameter and grip rods up to 19mm in diameter.
Ordering information: Comes with an extra set of fiberglass prong covers for temperatures above 100°C.